APO455837832 Apollo Scientific Ltd

2,3-Dichloro-4-fluoroaniline

  • MDL number MFCD08706409
  • Cas Number : 36556-52-2
  • Molecular Weight : 180.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-68
Synonym 1-Amino-2,3-dichloro-4-fluorobenzene
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/23-dichloro-4-fluoroaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/23-dichloro-4-fluoroaniline
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart