APO455837836 Apollo Scientific Ltd

Methyl 5-fluoro-2-nitrobenzoate

  • MDL number MFCD03425619
  • Cas Number : 393-85-1
  • Molecular Weight : 199.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35-37
Purity 98
Flash Point >110
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 115
Product Link https://store.apolloscientific.co.uk/product/methyl-5-fluoro-2-nitrobenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-5-fluoro-2-nitrobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart