APO455837843 Apollo Scientific Ltd

3-Amino-5-fluorobenzonitrile

  • MDL number MFCD05864357
  • Cas Number : 210992-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-107
Synonym 3-Cyano-5-fluoroaniline
Purity 97
Flash Point >110
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-amino-5-fluorobenzonitrile

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-5-fluorobenzonitrile
pictograms GHS06;GHS07
hstatements H301+H311+H331;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart