APO455837846 Apollo Scientific Ltd

5'-Bromo-2',4'-difluoroacetophenone

  • MDL number MFCD13190613
  • Cas Number : 864773-64-8
  • Molecular Weight : 235.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 31-32
Iupac Name 1-(5-bromo-2,4-difluorophenyl)ethanone
Inchi Code 1S/C8H5BrF2O/c1-4(12)5-2-6(9)8(11)3-7(5)10/h2-3H,1H3
Inchi Key KXZHDRBQPYKHKS-UHFFFAOYSA-N

Description

Synonym 1-(5-Bromo-2,4-difluorophenyl)ethan-1-one
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 31-32
Product Link https://store.apolloscientific.co.uk/product/5-bromo-24-difluoroacetophenone

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-24-difluoroacetophenone
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart