APO455837852 Apollo Scientific Ltd

3-Fluoro-4-[(2-hydroxyethyl)carbamoyl]benzeneboronic acid

  • MDL number MFCD08689535
  • Cas Number : 874289-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 217-219
Purity 98
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=35689

Safety Information

signalword Warning
pcodes P280;P271;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=35689
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart