APO455837859 Apollo Scientific Ltd

2-Fluoro-5-hydroxybenzonitrile

  • MDL number MFCD11846514
  • Cas Number : 104798-53-0
  • Molecular Weight : 137.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-108
Synonym 3-Cyano-4-fluorophenol
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-5-hydroxybenzonitrile

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-5-hydroxybenzonitrile
pictograms GHS06
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart