APO455837862 Apollo Scientific Ltd

5-Bromo-2-chloro-3-fluoropyridine

  • MDL number MFCD03411574
  • Cas Number : 831203-13-5
  • Molecular Weight : 210.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-52
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-chloro-3-fluoropyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-chloro-3-fluoropyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart