APO455837869 Apollo Scientific Ltd

4-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonyl chloride

  • MDL number MFCD07774162
  • Cas Number : 874838-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51-53
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=35980

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=35980
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart