APO455837879 Apollo Scientific Ltd

4-{[2-(Trifluoromethyl)phenyl]carbamoyl}benzeneboronic acid

  • MDL number MFCD09027216
  • Cas Number : 913835-42-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Borono-N-2-(trifluoromethyl)phenylbenzamide
Purity 98
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36029

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36029
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart