APO455837880 Apollo Scientific Ltd

Methyl 3-fluoro-4-nitrobenzoate

  • MDL number MFCD08444027
  • Cas Number : 185629-31-6
  • Molecular Weight : 199.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-54
Synonym 2-Fluoro-4-(methoxycarbonyl)nitrobenzene
Purity 98
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-3-fluoro-4-nitrobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-fluoro-4-nitrobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart