APO455837885 Apollo Scientific Ltd

2,4,5-Trifluoroaniline

  • MDL number MFCD00007649
  • Cas Number : 367-34-0
  • Molecular Weight : 147.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59-63
Purity 99
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/245-trifluoroaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/245-trifluoroaniline
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart