APO455837890 Apollo Scientific Ltd

1-(Difluoromethoxy)-2-iodobenzene

  • MDL number MFCD09749662
  • Cas Number : 920981-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Difluoromethyl 2-iodophenyl ether, alpha,alpha-Difluoro-2-iodoanisole
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36073

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=36073
pictograms GHS02;GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart