APO455837893 Apollo Scientific Ltd

2,3,6-Trifluorobenzaldehyde

  • MDL number MFCD00061195
  • Cas Number : 104451-70-9
  • Molecular Weight : 160.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.42
Flash Point 76
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 159
Product Link https://store.apolloscientific.co.uk/product/236-trifluorobenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/236-trifluorobenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart