APO455837895 Apollo Scientific Ltd

2,4,6-Trifluorobenzaldehyde

  • MDL number MFCD00061197
  • Cas Number : 58551-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-68
Purity 98
Density 1.407
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 165
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5004

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=5004
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart