APO455837904 Apollo Scientific Ltd

2H-Tridecafluoro-2-methylpentane

  • MDL number MFCD09763637
  • Cas Number : 30320-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 60-61
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36595

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=36595
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart