APO455837905 Apollo Scientific Ltd

4-Benzyloxy-2,3-difluorobenzeneboronic acid

  • MDL number MFCD07644469
  • Cas Number : 156635-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-benzyloxy-23-difluorobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/4-benzyloxy-23-difluorobenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart