APO455837912 Apollo Scientific Ltd

2,3,4-Trifluorobenzonitrile

  • MDL number MFCD00013288
  • Cas Number : 143879-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.12
Flash Point 68
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 116-118/100mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5027

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5027
pictograms GHS07
hstatements H302;H312;H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart