APO455837924 Apollo Scientific Ltd

2,3,4-Trifluorobenzyl alcohol

  • MDL number MFCD00083551
  • Cas Number : 144284-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 29-29.6
Purity 97
Physical Form Solid-Low Melt
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5046

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=5046
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart