APO455837928 Apollo Scientific Ltd

3,4,5-Trifluorobenzyl alcohol

  • MDL number MFCD00083527
  • Cas Number : 220227-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (3,4,5-Trifluorophenyl)methanol
Purity 97
Density 1.385
Flash Point 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 203-204
Product Link https://store.apolloscientific.co.uk/product/345-trifluorobenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/345-trifluorobenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart