APO455837931 Apollo Scientific Ltd

2,4,5-Trifluorobenzyl bromide

  • MDL number MFCD00061209
  • Cas Number : 157911-56-3
  • Molecular Weight : 225.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Bromomethyl)-2,4,5-trifluorobenzene, alpha-Bromo-2,4,5-trifluorotoluene
Purity 97
Density 1.71
Flash Point 90.6
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/245-trifluorobenzyl-bromide

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/245-trifluorobenzyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart