APO455837934 Apollo Scientific Ltd

1H,1H,10H,10H-Hexadecafluorodecane-1,10-diyl bis(2-propylpentanoate)

  • MDL number MFCD08059546
  • Cas Number : 544446-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluorodecane-1,10-diyl bis(2-propylpentanoate)
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10357

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10357
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart