APO455837945 Apollo Scientific Ltd

4-Fluoro-3-(trifluoromethoxy)benzonitrile

  • MDL number MFCD06660253
  • Cas Number : 886501-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Cyano-alpha,alpha,alpha,2-tetrafluoroanisole, 5-Cyano-2-fluorophenyl trifluoromethyl ether
Purity 98
Storage temp. Ambient Temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36694

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36694
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart