APO455837950 Apollo Scientific Ltd

4,4-Difluorocyclohexan-1-amine hydrochloride

  • MDL number MFCD06246881
  • Cas Number : 675112-70-6
  • Molecular Weight : 171.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 106-107
Iupac Name 4,4-difluorocyclohexanamine hydrochloride
Inchi Code 1S/C6H11F2N.ClH/c7-6(8)3-1-5(9)2-4-6;/h5H,1-4,9H2;1H
Inchi Key QYJPFTAKVBDDPD-UHFFFAOYSA-N

Description

Synonym 4-Amino-1,1-difluorocyclohexane hydrochloride
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 106-107
Product Link https://store.apolloscientific.co.uk/product/44-difluorocyclohexan-1-amine-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/44-difluorocyclohexan-1-amine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart