APO455837954 Apollo Scientific Ltd

4'-Fluoro-3'-methoxyacetophenone

  • MDL number MFCD00272135
  • Cas Number : 64287-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51-53
Purity 99
Flash Point 100
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3313

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=3313
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart