APO455837957 Apollo Scientific Ltd

6-Bromo-4-chloro-2-(trifluoromethyl)quinoline

  • MDL number MFCD00153073
  • Cas Number : 1701-28-6
  • Molecular Weight : 310.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-4-chloro-2-(trifluoromethyl)quinoline
Inchi Code 1S/C10H4BrClF3N/c11-5-1-2-8-6(3-5)7(12)4-9(16-8)10(13,14)15/h1-4H
Inchi Key JKEQQFAWABXNFV-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromo-4-chloro-2-trifluoromethylquinoline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromo-4-chloro-2-trifluoromethylquinoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart