APO455837962 Apollo Scientific Ltd

3-(Trifluoromethoxy)phenacyl bromide

  • MDL number MFCD03094425
  • Cas Number : 237386-01-5
  • Molecular Weight : 283.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-1-[3-(trifluoromethoxy)phenyl]ethanone
Inchi Code 1S/C9H6BrF3O2/c10-5-8(14)6-2-1-3-7(4-6)15-9(11,12)13/h1-4H,5H2
Inchi Key QUKYVVGHLJGVEQ-UHFFFAOYSA-N

Description

Synonym 2-Bromo-3'-(trifluoromethoxy)acetophenone, 2-Bromo-1-[3-(trifluoromethoxy)phenyl]ethan-1-one
Purity 97%
Flash Point 86
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-trifluoromethoxyphenacyl-bromide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-trifluoromethoxyphenacyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart