APO455837975 Apollo Scientific Ltd

3-Bromo-4-fluorobenzyl alcohol

  • MDL number MFCD00143093
  • Cas Number : 77771-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 80/0.5mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2468

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=2468
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart