APO455837981 Apollo Scientific Ltd

4-Iodo-1,1,1-trifluorobutane

  • MDL number MFCD00041543
  • Cas Number : 461-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4,4,4-Trifluorobutyl iodide
Purity 99
Density 1.85
Flash Point >100
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 126-127
Product Link https://store.apolloscientific.co.uk/product/4-iodo-111-trifluorobutane-99

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-iodo-111-trifluorobutane-99
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart