APO455837990 Apollo Scientific Ltd

2-Bromo-5-fluoro-4-methylaniline

  • MDL number MFCD00142871
  • Cas Number : 202865-78-9
  • Molecular Weight : 204.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Amino-5-bromo-2-fluorotoluene, 2-Bromo-5-fluoro-p-toluidine
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-fluoro-4-methylaniline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-fluoro-4-methylaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart