APO455837995 Apollo Scientific Ltd

5-Bromo-2-iodobenzotrifluoride

  • MDL number MFCD00672935
  • Cas Number : 364-12-5
  • Molecular Weight : 350.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-72
Synonym 4-Bromo-1-iodo-2-(trifluoromethyl)benzene, 5-Bromo-2-iodo-alpha,alpha,alpha-trifluorotoluene
Purity 98
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-iodobenzotrifluoride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-iodobenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart