APO455837997 Apollo Scientific Ltd

4-Fluoro-3-(trifluoromethoxy)benzoic acid

  • MDL number MFCD06660161
  • Cas Number : 886496-49-7
  • Molecular Weight : 224.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 86-90
Iupac Name 4-fluoro-3-(trifluoromethoxy)benzoic acid
Inchi Code 1S/C8H4F4O3/c9-5-2-1-4(7(13)14)3-6(5)15-8(10,11)12/h1-3H,(H,13,14)
Inchi Key PTPKGMXRSREZFB-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 86-90
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-3-trifluoromethoxybenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-3-trifluoromethoxybenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart