APO455838020 Apollo Scientific Ltd

3-(Trifluoromethoxy)benzonitrile

  • MDL number MFCD00040959
  • Cas Number : 52771-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.31
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5170

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=5170
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart