APO455838025 Apollo Scientific Ltd

2-(Trifluoromethoxy)benzylamine

  • MDL number MFCD00085176
  • Cas Number : 175205-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [2-(Trifluoromethoxy)phenyl]methylamine
Purity 95
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 77/12mm
Product Link https://store.apolloscientific.co.uk/product/2-trifluoromethoxybenzylamine-95

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-trifluoromethoxybenzylamine-95
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart