APO455838028 Apollo Scientific Ltd

2-(Trifluoromethoxy)benzyl alcohol

  • MDL number MFCD00153285
  • Cas Number : 175278-07-6
  • Molecular Weight : 192.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [2-(Trifluoromethoxy)phenyl]methanol
Purity 97
Density 1.33
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 96-98/12mm
Product Link https://store.apolloscientific.co.uk/product/2-trifluoromethoxybenzyl-alcohol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-trifluoromethoxybenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart