APO455838033 Apollo Scientific Ltd

2-(Trifluoromethoxy)benzyl chloride

  • MDL number MFCD00221456
  • Cas Number : 116827-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 68-70/15mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5186

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=5186
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart