APO455838034 Apollo Scientific Ltd

4-(Trifluoromethoxy)benzyl chloride

  • MDL number MFCD00052326
  • Cas Number : 65796-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.34
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 72-73/15mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5187

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=5187
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart