APO455838043 Apollo Scientific Ltd

3-(Trifluoromethoxy)phenol

  • MDL number MFCD00040987
  • Cas Number : 827-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym alpha,alpha,alpha-Trifluoro-m-guaiacol
Purity 98
Density 1.379
Flash Point 84
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 69-70/12mm
Product Link https://store.apolloscientific.co.uk/product/3-trifluoromethoxyphenol-98

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-trifluoromethoxyphenol-98
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart