APO455838046 Apollo Scientific Ltd

4-(Trifluoromethoxy)phenoxyacetic hydrazide

  • MDL number MFCD00052355
  • Cas Number : 175204-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133-135
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5213

Safety Information

signalword Warning
pcodes P280;P301+P312;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5213
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart