APO455838051 Apollo Scientific Ltd

4-(Trifluoromethoxy)phenylthiourea

  • MDL number MFCD00052992
  • Cas Number : 142229-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-140
Synonym 1-(Carbamothioylamino)-4-(trifluoromethoxy)benzene, 1-Thioureido-4-(trifluoromethoxy)benzene
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5237

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5237
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart