APO455838053 Apollo Scientific Ltd

4-(Trifluoromethoxy)thiophenol

  • MDL number MFCD00052312
  • Cas Number : 169685-29-4
  • Molecular Weight : 194.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(trifluoromethoxy)benzenethiol
Inchi Code 1S/C7H5F3OS/c8-7(9,10)11-5-1-3-6(12)4-2-5/h1-4,12H
Inchi Key JHVNSRJPBXPZJU-UHFFFAOYSA-N

Description

Synonym 4-(Trifluoromethoxy)benzenethiol, 4-(Trifluoromethoxy)phenyl mercaptan
Purity 95%
Flash Point 64-66/15mm
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-trifluoromethoxythiophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-trifluoromethoxythiophenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart