APO455838055 Apollo Scientific Ltd

3-(Trifluoromethoxy)phenylacetic acid

  • MDL number MFCD00082480
  • Cas Number : 203302-97-0
  • Molecular Weight : 220.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp >110
bp 53-55
Iupac Name [3-(trifluoromethoxy)phenyl]acetic acid
Inchi Code 1S/C9H7F3O3/c10-9(11,12)15-7-3-1-2-6(4-7)5-8(13)14/h1-4H,5H2,(H,13,14)
Inchi Key NFZQVADYFXRRPM-UHFFFAOYSA-N

Description

Melting Point >110
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 53-55
Product Link https://store.apolloscientific.co.uk/product/3-trifluoromethoxyphenylacetic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-trifluoromethoxyphenylacetic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart