APO455838057 Apollo Scientific Ltd

3-(Trifluoromethoxy)phenylacetonitrile

  • MDL number MFCD00061272
  • Cas Number : 108307-56-8
  • Molecular Weight : 201.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-trifluoromethoxyphenylacetonitrile

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-trifluoromethoxyphenylacetonitrile
pictograms GHS06;GHS07
hstatements H301;H312+H332;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart