APO455838060 Apollo Scientific Ltd

4'-(Trifluoromethyl)acetanilide

  • MDL number MFCD00013562
  • Cas Number : 349-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-154
Purity 98
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5246

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=5246
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart