APO455838062 Apollo Scientific Ltd

2-(Methylsulphonyl)-5-(trifluoromethyl)benzoic acid

  • MDL number MFCD09264536
  • Cas Number : 1000339-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176-178
Synonym 2-Carboxy-4-(trifluoromethyl)phenyl methyl sulphone, 3-Carboxy-4-(methylsulphonyl)benzotrifluoride
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36911

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36911
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart