APO455838064 Apollo Scientific Ltd

2-Fluoro-4-(methylsulphonyl)phenol

  • MDL number MFCD09264549
  • Cas Number : 398456-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-94
Synonym 3-Fluoro-4-hydroxyphenyl methyl sulphone
Purity 95
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36928

Safety Information

signalword Danger
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36928
pictograms GHS05;GHS07
hstatements H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart