APO455838070 Apollo Scientific Ltd

2-Amino-5-(trifluoromethyl)benzoic acid

  • MDL number MFCD01569537
  • Cas Number : 83265-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177
Synonym 4-Amino-3-carboxybenzotrifluoride, 2-Carboxy-4-(trifluoromethyl)aniline
Purity 98
Flash Point >110
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-amino-5-trifluoromethylbenzoic-acid-98

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-5-trifluoromethylbenzoic-acid-98
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart