APO455838072 Apollo Scientific Ltd

4-(Trifluoromethyl)benzamide

  • MDL number MFCD00007998
  • Cas Number : 1891-90-3
  • Molecular Weight : 189.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-186
Synonym 4-Carbamoylbenzotrifluoride
Purity 99
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-trifluoromethylbenzamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-trifluoromethylbenzamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart