APO455838078 Apollo Scientific Ltd

3-Fluoro-5-methylanisole

  • MDL number MFCD13185608
  • Cas Number : 160911-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47230

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=47230
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart