APO455838080 Apollo Scientific Ltd

4-(Methylsulphonyl)-2-(trifluoromethyl)benzyl bromide

  • MDL number MFCD16606069
  • Cas Number : 934557-65-0
  • Molecular Weight : 317.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-123
Synonym 2-(Bromomethyl)-5-(methylsulphonyl)benzotrifluoride, 1-(Bromomethyl)-4-(methylsulphonyl)-2-(trifluoromethyl)benzene, 4-(Bromomethyl)-3-(trifluoromethyl)phenyl methyl sulphone
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-methylsulphonyl-2-trifluoromethylbenzyl-bromide

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-methylsulphonyl-2-trifluoromethylbenzyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart