APO455838088 Apollo Scientific Ltd

2-(Trifluoromethyl)benzonitrile

  • MDL number MFCD00001791
  • Cas Number : 447-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 17-19
Purity 98
Density 1.294
Flash Point 90
Physical Form Solid-Low Melt
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 204-206
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5282

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5282
pictograms GHS07
hstatements H302;H312;H332;H319;H412
Availability details for
Quantity
  • Availability
  • Add to Cart